Assay Details
Gene Symbol: F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024640.1 | NP_001019811.1 | 4 - 5 | 657 | 137 | |
XM_005621318.3 | XP_005621375.1 | 4 - 5 | 732 | 137 | ||
GenBank mRNA | AB200288.1 | - | 4 - 5 | 657 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
