Assay Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003168.1 | NP_001003168.1 | 69 | 111 | ||
GenBank mRNA | AF069071.1 | - | 69 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001003168.1 transcripts.
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
