Assay Details
Gene Symbol: RPE65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003176.1 | NP_001003176.1 | 8 - 9 | 886 | 122 | |
GenBank mRNA | AF084537.1 | - | 8 - 9 | 886 | 122 | |
Y16567.1 | - | 886 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
