Assay Details
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003305.1 | NP_001003305.1 | 1 - 2 | 187 | 74 | |
GenBank mRNA | AF091131.1 | - | 1 - 2 | 98 | 74 | |
AF333117.1 | - | 1 - 2 | 130 | 74 | ||
AF333118.1 | - | 1 - 2 | 49 | 74 | ||
D30710.1 | - | 1 - 2 | 180 | 74 | ||
U11689.1 | - | 1 - 2 | 149 | 74 | ||
U28141.1 | - | 1 - 2 | 187 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
