Assay Details
Gene Symbol: IL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006950.1 | NP_001006951.1 | 2 - 3 | 211 | 101 | |
GenBank mRNA | AF091133.1 | - | 2 - 3 | 147 | 101 | |
AF331919.1 | - | 2 - 3 | 211 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
