Assay Details
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003384.1 | NP_001003384.1 | 1 - 2 | 177 | 76 | |
XM_022424906.1 | XP_022280614.1 | 1 - 2 | 771 | 76 | ||
GenBank mRNA | AF244915.1 | - | 1 - 2 | 177 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
