Assay Details
Gene Symbol: PPP2CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003063.1 | NP_001003063.1 | 2 - 3 | 316 | 145 | |
GenBank mRNA | AF448499.1 | - | 2 - 3 | 316 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() |
