Assay Details
Gene Symbol: PPP1CC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003033.1 | NP_001003033.1 | 186 | 113 | ||
XM_005636001.3 | XP_005636058.1 | 464 | 113 | |||
GenBank mRNA | AF525130.1 | - | 186 | 113 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
