Assay Details
Gene Symbol: PRKACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003032.1 | NP_001003032.1 | 2 - 3 | 237 | 66 | |
GenBank mRNA | AF525132.1 | - | 2 - 3 | 237 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
