Assay Details
Gene Symbol: UBE2S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286964.1 | NP_001273893.1 | 4 | 831 | 95 | |
GenBank mRNA | AJ388535.1 | - | 1 | 144 | 95 | |
GR889260.1 | - | 4 | 705 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
