Assay Details
Gene Symbol: RABAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003014.1 | NP_001003014.1 | 1 - 2 | 131 | 82 | |
GenBank mRNA | AJ534245.1 | - | 1 - 2 | 131 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
