Assay Details
Gene Symbol: TACR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012619.1 | NP_001012637.1 | 4 - 5 | 933 | 61 | |
GenBank mRNA | AJ884915.1 | - | 4 - 5 | 933 | 61 | |
S75109.1 | - | 1 - 2 | 146 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
