Assay Details
Gene Symbol: BHLHE41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002973.1 | NP_001002973.1 | 507 | 120 | ||
GenBank mRNA | AY204568.1 | - | 507 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
