Assay Details
Gene Symbol: MITF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003337.1 | NP_001003337.1 | 696 | 73 | ||
XM_005632016.2 | XP_005632073.1 | 911 | 73 | |||
GenBank mRNA | AY240952.1 | - | 696 | 73 |
