Assay Details
Gene Symbol: CAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003359.1 | NP_001003359.1 | 1 - 2 | 237 | 61 | |
GenBank mRNA | AY392089.1 | - | 1 - 2 | 237 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
