Assay Details
Gene Symbol: GNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003364.1 | NP_001003364.1 | 1 - 2 | 157 | 56 | |
GenBank mRNA | AY563036.1 | - | 1 - 2 | 157 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
