Assay Details
Gene Symbol: UOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011886.1 | NP_001011886.1 | 7 - 8 | 869 | 149 | |
GenBank mRNA | AY871313.1 | - | 7 - 8 | 869 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
