Assay Details
Gene Symbol: FCER1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110766.2 | NP_001104236.2 | 1 - 2 | 51 | 80 | |
GenBank mRNA | D16413.1 | - | 1 - 2 | 59 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
