Assay Details
Gene Symbol: FCER1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110766.2 | NP_001104236.2 | 4 - 5 | 598 | 73 | |
XM_022415289.1 | XP_022270997.1 | 4 - 5 | 654 | 73 | ||
GenBank mRNA | D16413.1 | - | 4 - 5 | 606 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
