Assay Details
Gene Symbol: GUCY1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018034.2 | NP_001018044.1 | 10 - 11 | 1573 | 74 | |
GenBank mRNA | DQ008576.1 | - | 10 - 11 | 1573 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
