Assay Details
Gene Symbol: RAB5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003317.2 | NP_001003317.1 | 3 - 4 | 439 | 87 | |
XM_022408381.1 | XP_022264089.1 | 3 - 4 | 475 | 87 | ||
GenBank mRNA | M35520.1 | - | 3 - 4 | 439 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
