Assay Details
Gene Symbol: PDE6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003235.1 | NP_001003235.1 | 37 | 142 | ||
GenBank mRNA | U49359.1 | - | 37 | 142 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001003235.1 transcripts.
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
