Assay Details
Gene Symbol: SRP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003251.1 | NP_001003251.1 | 3 - 4 | 214 | 135 | |
GenBank mRNA | U57440.1 | - | 3 - 4 | 214 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
