Assay Details
Gene Symbol: G6PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002993.2 | NP_001002993.2 | 402 | 78 | ||
XM_022422515.1 | XP_022278223.1 | 402 | 78 | |||
GenBank mRNA | U91844.2 | - | 402 | 78 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
