Assay Details
Gene Symbol: SLC16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_533065.5 | XP_533065.2 | 1 - 2 | 134 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_533065.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
