Assay Details
Gene Symbol: INTS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197314.1 | NP_001184243.1 | 5 - 6 | 689 | 73 | |
XM_005636713.3 | XP_005636770.1 | 6 - 7 | 1241 | 73 | ||
XM_005636714.3 | XP_005636771.1 | 6 - 7 | 827 | 73 | ||
XM_022410862.1 | XP_022266570.1 | 6 - 7 | 1241 | 73 | ||
XM_022410863.1 | XP_022266571.1 | 5 - 6 | 1005 | 73 | ||
XM_022410864.1 | XP_022266572.1 | 4 - 5 | 930 | 73 | ||
GenBank mRNA | GU811149.1 | - | 5 - 6 | 689 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
