Assay Details
Gene Symbol: INTS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197314.1 | NP_001184243.1 | 6 - 7 | 813 | 127 | |
XM_005636713.3 | XP_005636770.1 | 7 - 8 | 1365 | 127 | ||
XM_005636714.3 | XP_005636771.1 | 7 - 8 | 951 | 127 | ||
XM_022410862.1 | XP_022266570.1 | 7 - 8 | 1365 | 127 | ||
XM_022410863.1 | XP_022266571.1 | 6 - 7 | 1129 | 127 | ||
XM_022410864.1 | XP_022266572.1 | 5 - 6 | 1054 | 127 | ||
GenBank mRNA | GU811149.1 | - | 6 - 7 | 813 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() |
