Assay Details
Gene Symbol: TPPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629145.3 | XP_005629202.1 | 2 - 3 | 352 | 82 | |
XM_005629146.3 | XP_005629203.1 | 2 - 3 | 690 | 82 | ||
XM_022428092.1 | XP_022283800.1 | 1 - 2 | 233 | 82 | ||
XM_022428093.1 | XP_022283801.1 | 1 - 2 | 236 | 82 | ||
XM_022428094.1 | XP_022283802.1 | 1 - 2 | 235 | 82 | ||
XM_022428095.1 | XP_022283803.1 | 1 - 2 | 235 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
