Assay Details
Gene Symbol: PTOV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616289.3 | XP_005616346.1 | 9 - 10 | 1094 | 128 | |
XM_846350.5 | XP_851443.1 | 9 - 10 | 1094 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
