Assay Details
Gene Symbol: STON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204842.1 | NP_001191771.1 | 1 - 2 | 1936 | 93 | |
XM_005625666.2 | XP_005625723.1 | 3 - 4 | 2174 | 93 | ||
XM_005625667.3 | XP_005625724.1 | 2 - 3 | 2052 | 93 | ||
GenBank mRNA | DN905073.1 | - | 1 - 2 | 510 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
