Assay Details
Gene Symbol: MAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005633484.3 | XP_005633541.1 | 1 - 2 | 1588 | 88 | |
XM_022407675.1 | XP_022263383.1 | 1 - 2 | 1895 | 88 | ||
XM_534007.6 | XP_534007.3 | 1 - 2 | 1588 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
