Assay Details
Gene Symbol: LOC608697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014106509.2 | XP_013961984.1 | 4 - 5 | 362 | 85 | |
XM_014106515.2 | XP_013961990.1 | 4 - 5 | 362 | 85 | ||
XM_022408603.1 | XP_022264311.1 | 4 - 5 | 362 | 85 | ||
XM_022408604.1 | XP_022264312.1 | 4 - 5 | 362 | 85 | ||
XM_022408607.1 | XP_022264315.1 | 3 - 4 | 285 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022408607.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
