Assay Details
Gene Symbol: CUTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637608.3 | XP_005637665.1 | 3 - 4 | 347 | 97 | |
XM_014109040.2 | XP_013964515.1 | 3 - 4 | 484 | 97 | ||
XM_014109041.2 | XP_013964516.1 | 3 - 4 | 289 | 97 | ||
XM_022412122.1 | XP_022267830.1 | 3 - 4 | 347 | 97 | ||
XM_534987.6 | XP_534987.2 | 3 - 4 | 347 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
