Assay Details
Gene Symbol: COMMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252346.1 | NP_001239275.1 | 546 | 93 | ||
XM_005616928.2 | XP_005616985.1 | 549 | 93 | |||
XM_005616929.3 | XP_005616986.1 | 477 | 93 | |||
GenBank mRNA | DN751136.1 | - | 369 | 93 | ||
DN905434.1 | - | 546 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
