Assay Details
Gene Symbol: PMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640441.3 | XP_005640498.1 | 4 - 5 | 814 | 156 | |
XM_005640442.3 | XP_005640499.1 | 4 - 5 | 572 | 156 | ||
XM_005640443.3 | XP_005640500.1 | 4 - 5 | 562 | 156 | ||
XM_005640444.3 | XP_005640501.1 | 4 - 5 | 454 | 156 | ||
XM_005640447.3 | XP_005640504.1 | 3 - 4 | 312 | 156 | ||
XM_005640449.3 | XP_005640506.1 | 4 - 5 | 495 | 156 | ||
XM_014110946.2 | XP_013966421.1 | 4 - 5 | 459 | 156 | ||
XM_014110947.2 | XP_013966422.1 | 5 - 6 | 1037 | 156 | ||
XM_022414962.1 | XP_022270670.1 | 4 - 5 | 495 | 156 | ||
XM_022414963.1 | XP_022270671.1 | 4 - 5 | 495 | 156 | ||
XM_022414964.1 | XP_022270672.1 | 4 - 5 | 495 | 156 | ||
XM_022414965.1 | XP_022270673.1 | 4 - 5 | 495 | 156 | ||
XM_536002.6 | XP_536002.1 | 4 - 5 | 495 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
