Assay Details
Gene Symbol: ZNF567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014115506.2 | XP_013970981.1 | 4 - 5 | 602 | 76 | |
XM_022403730.1 | XP_022259438.1 | 4 - 5 | 359 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014115506.2, XM_022403730.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
