Assay Details
Gene Symbol: RTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635200.3 | XP_005635257.1 | 1 - 2 | 155 | 72 | |
XM_014107159.2 | XP_013962634.1 | 1 - 2 | 155 | 72 | ||
XM_848341.5 | XP_853434.1 | 1 - 2 | 155 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
