Assay Details
Gene Symbol: RPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111641.2 | XP_013967116.1 | 3 - 4 | 334 | 92 | |
XM_014111642.2 | XP_013967117.1 | 3 - 4 | 336 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
