Assay Details
Gene Symbol: IKBIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629259.3 | XP_005629316.1 | 1 - 2 | 493 | 150 | |
XM_539736.6 | XP_539736.2 | 1 - 2 | 500 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
