Assay Details
Gene Symbol: WBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022404798.1 | XP_022260506.1 | 4 - 5 | 931 | 100 | |
XM_847547.5 | XP_852640.1 | 4 - 5 | 943 | 100 | ||
XM_860965.5 | XP_866058.2 | 3 - 4 | 840 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
