Assay Details
Gene Symbol: LMAN2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022424658.1 | XP_022280366.1 | 4 - 5 | 514 | 67 | |
XM_022424659.1 | XP_022280367.1 | 4 - 5 | 618 | 67 | ||
XM_531797.6 | XP_531797.4 | 7 - 8 | 852 | 67 | ||
XM_858841.5 | XP_863934.3 | 6 - 7 | 817 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
