Assay Details
Gene Symbol: BICRAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627362.3 | XP_005627419.1 | 8 - 9 | 2230 | 91 | |
XM_005627363.3 | XP_005627420.1 | 8 - 9 | 2214 | 91 | ||
XM_014118453.2 | XP_013973928.1 | 8 - 9 | 2547 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
