Assay Details
Gene Symbol: JAZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022427520.1 | XP_022283228.1 | 3 - 4 | 435 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: JAZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022427520.1 | XP_022283228.1 | 3 - 4 | 435 | 81 |
Set Membership: |
![]() ![]() ![]() ![]() |