Assay Details
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022422460.1 | XP_022278168.1 | 5 - 6 | 950 | 149 | |
XM_022422461.1 | XP_022278169.1 | 6 - 7 | 1015 | 149 | ||
XM_533564.6 | XP_533564.3 | 6 - 7 | 1012 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() |
