Assay Details
Gene Symbol: PLAAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631586.2 | XP_005631643.1 | 2 - 3 | 224 | 79 | |
XM_533254.6 | XP_533254.3 | 2 - 3 | 224 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
