Assay Details
Gene Symbol: SERPINB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615344.3 | XP_005615401.1 | 434 | 73 | ||
XM_022414958.1 | XP_022270666.1 | 754 | 73 | |||
XM_022415006.1 | XP_022270714.1 | 529 | 73 | |||
XM_533382.5 | XP_533382.3 | 676 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
