Assay Details
Gene Symbol: HSD17B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047981.1 | NP_001041446.1 | 5 - 6 | 416 | 95 | |
XM_005615748.3 | XP_005615805.1 | 5 - 6 | 520 | 95 | ||
XM_014114134.2 | XP_013969609.1 | 4 - 5 | 427 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
