Assay Details
Gene Symbol: GRINA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426865.1 | XP_022282573.1 | 5 - 6 | 811 | 58 | |
XM_532348.6 | XP_532348.3 | 4 - 5 | 778 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
