Assay Details
Gene Symbol: RIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426550.1 | XP_022282258.1 | 1 | 304 | 88 | |
XM_532278.6 | XP_532278.4 | 1 | 303 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
