Assay Details
Gene Symbol: MRPL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426086.1 | XP_022281794.1 | 2 - 3 | 107 | 82 | |
XM_532153.6 | XP_532153.1 | 2 - 3 | 186 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
